siRNA Details
Gene Symbol: 1810059H22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079377.1 | - | 4 | 847 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE332870.1 | - | 1 | 241 | Chr. 5 - 148954319 - 148954668 [-] on Build GRCm38 |
