siRNA Details
Gene Symbol: Gm26910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078979.1 | - | 2 | 574 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV379027.2 | - | 2 | 544 | Chr. 18 - 67773713 - 67774828 [-] on Build GRCm38 |
