siRNA Details
Gene Symbol: 1700039E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045315.1 | 3 | 718 | |||
GenBank mRNA | AK006641.1 | - | 3 | 722 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700039E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045315.1 | 3 | 718 | |||
GenBank mRNA | AK006641.1 | - | 3 | 722 |