siRNA Details
Gene Symbol: 1700039E22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045315.1 | 2, 3 | 575 | |||
GenBank mRNA | AK006641.1 | - | 2, 3 | 579 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706631.1 | - | 2, 3 | 582 | Chr. 19 - 44828493 - 44835934 [-] on Build GRCm38 |
