siRNA Details
Gene Symbol: Gm15614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136552.1 | - | 4 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ054039.1 | - | 4 | 503 | Chr. 5 - 32508099 - 32583386 [-] on Build GRCm38 |
