siRNA Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030803.1 | - | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB619731.1 | - | 3 | 527 | Chr. 6 - 125265476 - 125269312 [+] on Build GRCm38 |
