siRNA Details
Gene Symbol: A930019D19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040619.1 | 4 | 395 | |||
GenBank mRNA | AK020875.1 | - | 4 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742222.1 | - | 4 | 398 | Chr. 2 - 146259286 - 146266866 [+] on Build GRCm38 |
