siRNA Details
Gene Symbol: Gm20619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015224.1 | - | 4 | 635 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714779.1 | - | 4 | 635 | Chr. 6 - 6248690 - 6377625 [+] on Build GRCm38 |
