siRNA Details
Gene Symbol: 4930509E16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045735.1 | 3 | 202 | |||
GenBank mRNA | AK015733.1 | - | 3 | 202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715275.1 | - | 3 | 199 | Chr. 9 - 72518525 - 72531856 [-] on Build GRCm38 |
