siRNA Details
Gene Symbol: 4930401C12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015031.1 | - | 1 | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU939059.1 | - | 1 | 44 | Chr. 11 - 112156866 - 112157336 [-] on Build GRCm38 |
