siRNA Details
Gene Symbol: Gm16006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042803.1 | - | 2 | 181 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640009.1 | - | 2 | 181 | Chr. 15 - 57708203 - 57708927 [-] on Build GRCm38 |
