siRNA Details
Gene Symbol: Gm4872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073372.1 | 1 | 28 | |||
GenBank mRNA | AK043311.1 | - | 1 | 29 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB640765.1 | - | 1 | 27 | Chr. 6 - 53221803 - 53226601 [+] on Build GRCm38 |
