siRNA Details
Gene Symbol: 2610035F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045046.1 | 5 | 1044 | |||
GenBank mRNA | AK011684.1 | - | 5 | 1045 | ||
AK042842.1 | - | 3 | 1703 | |||
BB258337.1 | - | 2 | 249 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610035F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045046.1 | 5 | 1044 | |||
GenBank mRNA | AK011684.1 | - | 5 | 1045 | ||
AK042842.1 | - | 3 | 1703 | |||
BB258337.1 | - | 2 | 249 |