siRNA Details
Gene Symbol: 2610035F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045046.1 | 4 | 786 | |||
GenBank mRNA | AK011684.1 | - | 4 | 787 | ||
AK042842.1 | - | 3 | 643 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2610035F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045046.1 | 4 | 786 | |||
GenBank mRNA | AK011684.1 | - | 4 | 787 | ||
AK042842.1 | - | 3 | 643 |