siRNA Details
Gene Symbol: D430019H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052429.1 | - | 2 | 3044 | ||
AK081558.1 | - | 1 | 118 | |||
BC058677.1 | - | 1 | 2361 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D430019H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052429.1 | - | 2 | 3044 | ||
AK081558.1 | - | 1 | 118 | |||
BC058677.1 | - | 1 | 2361 |