siRNA Details
Gene Symbol: 9130025I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018650.1 | - | 1 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720835.1 | - | 1 | 390 | Chr. 1 - 159289061 - 159289807 [+] on Build GRCm38 |
