siRNA Details
Gene Symbol: 2900054C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013683.1 | - | 1 | 739 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ133100.1 | - | 1 | 499 | Chr. 17 - 90173129 - 90173776 [-] on Build GRCm38 |
