siRNA Details
Gene Symbol: 2210404E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008826.1 | - | 2 | 203 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708724.1 | - | 2 | 203 | Chr. 19 - 10684793 - 10685477 [+] on Build GRCm38 |
