siRNA Details
Gene Symbol: Gm8273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084737.1 | - | 1 | 468 | ||
BB479211.2 | - | 1 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB659405.1 | - | 1 | 463 | Chr. 4 - 8646957 - 8647448 [-] on Build GRCm38 |
