siRNA Details
Gene Symbol: C230079O03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040459.1 | 3 | 264 | |||
GenBank mRNA | AK048899.1 | - | 3 | 265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB651410.1 | - | 3 | 265 | Chr. 7 - 136332993 - 136349415 [-] on Build GRCm38 |
