siRNA Details
Gene Symbol: E130118H10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021395.1 | - | 1 | 146 | ||
AK165039.1 | - | 1 | 5310 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY731372.1 | - | 1 | 144 | Chr. 5 - 31572389 - 31573011 [+] on Build GRCm38 |
