siRNA Details
Gene Symbol: E130119H09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021396.1 | - | 1 | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY731386.1 | - | 1 | 233 | Chr. 13 - 53912644 - 53915748 [-] on Build GRCm38 |
