siRNA Details
Gene Symbol: Gm30371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC072646.1 | - | 3 | 556 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF532906.1 | - | 3 | 557 | Chr. 16 - 8944913 - 8966154 [+] on Build GRCm38 |
