siRNA Details
Gene Symbol: 5031439G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030332.1 | - | 3305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI649965.1 | - | 1 | 30 | Chr. 15 - 84944414 - 84945220 [-] on Build GRCm38 |
