siRNA Details
Gene Symbol: 1700007F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040538.1 | 2, 3 | 91 | |||
GenBank mRNA | AK005707.1 | - | 2, 3 | 72 | ||
AK017083.1 | - | 2, 3 | 93 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716761.1 | - | 2, 3 | 93 | Chr. 3 - 58142652 - 58163809 [-] on Build GRCm38 |
