siRNA Details
Gene Symbol: 2600014E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011210.1 | - | 1 | 725 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DR754642.1 | - | 1 | 253 | Chr. 2 - 75315635 - 75316154 [+] on Build GRCm38 |
