siRNA Details
Gene Symbol: Foxo6os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028616.1 | - | 1 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB612793.1 | - | 1 | 294 | Chr. 4 - 120291350 - 120302226 [+] on Build GRCm38 |
