siRNA Details
Gene Symbol: Gm1720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046015.1 | 7 | 495 | |||
GenBank mRNA | AK144520.1 | - | 7 | 496 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY733817.1 | - | 7 | 496 | Chr. X - 166662025 - 166683302 [+] on Build GRCm38 |
