siRNA Details
Gene Symbol: 4930513D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045416.1 | 5 | 595 | |||
GenBank mRNA | AK015779.1 | - | 5 | 595 | ||
AK132902.1 | - | 5 | 554 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB615423.1 | - | 5 | 554 | Chr. 5 - 39462735 - 39603531 [-] on Build GRCm38 |
