siRNA Details
Gene Symbol: 4930532M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108050.1 | 3 | 451 | |||
GenBank mRNA | AK015950.1 | - | 3 | 453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715514.1 | - | 3 | 452 | Chr. 1 - 154255689 - 154270418 [+] on Build GRCm38 |
