siRNA Details
Gene Symbol: 4930578E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045391.1 | 3 | 949 | |||
GenBank mRNA | AK019814.1 | - | 2 | 319 | ||
AK132929.1 | - | 3 | 950 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU605115.1 | - | 3 | 365 | Chr. 18 - 29451745 - 29495832 [-] on Build GRCm38 |
