siRNA Details
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009838.1 | - | 1 | 1887 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI686571.1 | - | 1 | 322 | Chr. 15 - 98038778 - 98039625 [-] on Build GRCm38 |
