siRNA Details
Gene Symbol: Gm15941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045283.1 | 3 | 846 | |||
GenBank mRNA | AK135939.1 | - | 3 | 847 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV649172.1 | - | 2 | 584 | Chr. 15 - 37422046 - 37427438 [-] on Build GRCm38 |
