siRNA Details
Gene Symbol: Gm15941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045283.1 | 1 | 21 | |||
GenBank mRNA | AK135939.1 | - | 1 | 22 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY719870.1 | - | 1 | 19 | Chr. 15 - 37422273 - 37432662 [-] on Build GRCm38 |
