siRNA Details
Gene Symbol: 4933413C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016799.1 | - | 2 | 498 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716475.1 | - | 2 | 498 | Chr. 19 - 28581824 - 28583294 [-] on Build GRCm38 |
