siRNA Details
Gene Symbol: C030018K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045411.1 | 3 | 492 | |||
GenBank mRNA | AK021093.1 | - | 3 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728189.1 | - | 3 | 486 | Chr. 5 - 64477015 - 64479572 [+] on Build GRCm38 |
