siRNA Details
Gene Symbol: C030018G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021090.1 | - | 1 | 79 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728185.1 | - | 1 | 77 | Chr. 10 - 108685129 - 108685793 [-] on Build GRCm38 |
