siRNA Details
Gene Symbol: BC058420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052319.1 | - | 1 | 2807 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF744037.1 | - | 1 | 350 | Chr. 7 - 131990789 - 131991510 [+] on Build GRCm38 |
