siRNA Details
Gene Symbol: 8430436N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040645.1 | 3 | 833 | |||
GenBank mRNA | AK018465.1 | - | 3 | 834 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8430436N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040645.1 | 3 | 833 | |||
GenBank mRNA | AK018465.1 | - | 3 | 834 |