siRNA Details
Gene Symbol: 9130208D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020274.1 | - | 1 | 513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720846.1 | - | 1 | 513 | Chr. 7 - 105823251 - 105823806 [-] on Build GRCm38 |
