siRNA Details
Gene Symbol: 3110067G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014239.1 | - | 1 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713199.1 | - | 1 | 140 | Chr. 3 - 152483734 - 152484403 [-] on Build GRCm38 |
