siRNA Details
Gene Symbol: 1110035E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004108.1 | - | 1 | 199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY704351.1 | - | 1 | 199 | Chr. 8 - 107312677 - 107313671 [+] on Build GRCm38 |
