siRNA Details
Gene Symbol: A930012N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020850.1 | - | 1 | 275 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742182.1 | - | 1 | 256 | Chr. 2 - 156180595 - 156181113 [+] on Build GRCm38 |
