siRNA Details
Gene Symbol: A930014D07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020854.1 | - | 2 | 312 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY742188.1 | - | 2 | 312 | Chr. 13 - 99007099 - 99021380 [+] on Build GRCm38 |
