siRNA Details
Gene Symbol: Gm5427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076335.1 | - | 170 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB612635.1 | - | 2 | 170 | Chr. 10 - 98655706 - 98699710 [-] on Build GRCm38 |
