siRNA Details
Gene Symbol: 4933433G15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040719.1 | 1 | 387 | |||
GenBank mRNA | AK014947.1 | - | 1 | 389 | ||
AK017040.1 | - | 388 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714464.1 | - | 1 | 383 | Chr. 9 - 75410188 - 75414377 [+] on Build GRCm38 |
