siRNA Details
Gene Symbol: Gm20740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041852.1 | - | 1 | 3768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB233142.2 | - | 1 | 278 | Chr. 15 - 63445126 - 63445730 [-] on Build GRCm38 |
