siRNA Details
Gene Symbol: 9030407P20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018490.1 | - | 2 | 318 | ||
AK034016.1 | - | 3 | 527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625892.1 | - | 3 | 522 | Chr. 7 - 120675088 - 120677515 [-] on Build GRCm38 |
