siRNA Details
Gene Symbol: 4930401B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089549.1 | - | 1 | 1273 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY547435.1 | - | 1 | 294 | Chr. 1 - 86435670 - 86436036 [+] on Build GRCm38 |
