siRNA Details
Gene Symbol: Gm6307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045331.1 | 4 | 1120 | |||
GenBank mRNA | AK140896.1 | - | 4 | 1123 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX729663.1 | - | 3 | 363 | Chr. 2 - 180386203 - 180401144 [+] on Build GRCm38 |
